Learn More
Biochain Institute Inc Total Protein Western Blots - Human Adult Normal Tissue, Blot IV, 16 lanes
SDP
Catalog No. 50183838
Encompass_Preferred
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. Patient demographic and disease related history data are available. Total Protein Western blots are manufactured using high quality proteins from the tissue repository. Proteins are isolated from these tissues using BioChain's proprietary techniques. The proteins are separated by 4-20% gradient SDS-PAGE gel electrophoresis, and transferred onto a PVDF membrane. Human placenta protein is used as an interblot control in each blot. A 10 kd to 250 kd pre-stained protein marker can be visualized on each blot. These blots are suitable for use in both radioactive and non-radioactive detection. They are supplied ready for use so that you can immediately screen the protein of interest without the hassle of obtaining hard to find tissues and processing the proteins yourself.
Catalog No. 50-183-838 Supplier Biochain Institute Inc Supplier No. W1234404
BioChain's tissue products are based on the sample repository network established following the IRB-approved ethical standard and procedures. Patient demographic and disease related history data are available. Total Protein Western blots are manufactured using high quality proteins from the tissue repository. Proteins are isolated from these tissues using BioChain's proprietary techniques. The proteins are separated by 4-20% gradient SDS-PAGE gel electrophoresis, and transferred onto a PVDF membrane. Human placenta protein is used as an interblot control in each blot. A 10 kd to 250 kd pre-stained protein marker can be visualized on each blot. These blots are suitable for use in both radioactive and non-radioactive detection. They are supplied ready for use so that you can immediately screen the protein of interest without the hassle of obtaining hard to find tissues and processing the proteins yourself.