Learn More
Cell Signaling Technology Total PSA/KLK3 Matched Antibody Pair, Unconjugated, Human, 1 Kit
SDP
Catalog No. 503134245
Encompass
This matched antibody pair is designed for immunoassay technologies and high-throughput ELISA platforms, including those requiring custom conjugation such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The kit includes capture and detection antibodies that bind to non-overlapping epitopes for optimal sandwich immunoassay performance.
  • Compatible with custom labeling including fluorophores, lanthanides, biotin, and beads
  • Supplied in BSA and azide-free formulation
  • Validated for human reactivity
  • Components consist of two rabbit monoclonal antibodies
  • Requires storage at -20°C
Catalog No. 50-313-4245 Supplier Cell Signaling Technology Supplier No. 76325S
This matched antibody pair is designed for immunoassay technologies and high-throughput ELISA platforms, including those requiring custom conjugation such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The kit includes capture and detection antibodies that bind to non-overlapping epitopes for optimal sandwich immunoassay performance.
  • Compatible with custom labeling including fluorophores, lanthanides, biotin, and beads
  • Supplied in BSA and azide-free formulation
  • Validated for human reactivity
  • Components consist of two rabbit monoclonal antibodies
  • Requires storage at -20°C