Learn More
ERA WATERS CORP TOTAL RESID. CHLORINE PT-WP225
SDP
Catalog No. NC0566943
Encompass
One 2 mL flame-sealed ampule yields up to 2 liters after dilution. Use with titrimetric or colorimetric methods.
Catalog No. NC0566943 Supplier ERA WATERS CORP Supplier No. 587
One 2 mL flame-sealed ampule yields up to 2 liters after dilution. Use with titrimetric or colorimetric methods.