Learn More
Total Residual Chlorine Drinking Water/Wastewater Standard (ULTRAcheck™ Blind)
Catalog No. USQCI780
Encompass
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
US-QCI-780 10 mL
1 options
Catalog No. US-QCI-780 Supplier Agilent Technologies Supplier No. QCI780
Total Residual Chlorine at 0.3-5.0mg/L when diluted to 2 liters
Specifications
Chemical Name or Material Total Residual Chlorine Drinking Water/Wastewater Standard
Packaging Ampule
Name Note ULTRAcheck Blind QC Standard
Quantity 10 mL