Learn More
Biochain Institute Inc Total RNA - Human Adult Normal Tissue: Brain: Cerebral Cortex, Ambient temperature shipping, 10 ug/pk
SDP
Catalog No. 501871503
Encompass_Preferred
Total RNA is isolated by modified guanidine thiocyanate techniques and stored in RNA storage buffer. The vials were then dried for room temperature shipping and storage. The total RNA need to be reconsitituted before use. Store the vials at -80 ℃ after reconstitution.
Catalog No. 50-187-1503 Supplier Biochain Institute Inc Supplier No. ATR123404210
Total RNA is isolated by modified guanidine thiocyanate techniques and stored in RNA storage buffer. The vials were then dried for room temperature shipping and storage. The total RNA need to be reconsitituted before use. Store the vials at -80 ℃ after reconstitution.