Learn More
Biochain Institute Inc Total RNA - Human Adult Normal Tissue: Kidney, 50 ug/PK
SDP
Catalog No. 50183940
Encompass_Preferred
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA polysaccharides, and proteoglycans has been effectively eliminated. The integrity of the total RNAs is assured by checking for a ratio of greater than 1:1 between 28s and 18s ribosomal RNA. High efficiency reverse transcription using the Total RNA is demonstrated.
Catalog No. 50-183-940 Supplier Biochain Institute Inc Supplier No. R123414250
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA polysaccharides, and proteoglycans has been effectively eliminated. The integrity of the total RNAs is assured by checking for a ratio of greater than 1:1 between 28s and 18s ribosomal RNA. High efficiency reverse transcription using the Total RNA is demonstrated.