Learn More
Biochain Institute Inc Total RNA - Alzheimer's Disease: Parietal Lobe, 10 ug/PK
SDP
Numéro de catalogue. 50183990
Encompass_Preferred
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA polysaccharides, and proteoglycans has been effectively eliminated. The integrity of the total RNAs is assured by checking for a ratio of greater than 1:1 between 28s and 18s ribosomal RNA. High efficiency reverse transcription using the Total RNA is demonstrated.
Numéro de catalogue. 50-183-990 Fournisseur Biochain Institute Inc Code fournisseur R1236066ALZ10
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA polysaccharides, and proteoglycans has been effectively eliminated. The integrity of the total RNAs is assured by checking for a ratio of greater than 1:1 between 28s and 18s ribosomal RNA. High efficiency reverse transcription using the Total RNA is demonstrated.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.