Learn More
Biochain Institute Inc Total RNA - Human Tumor Tissue: Rectum, 50 ug/PK
SDP
Catalog No. 50184069
Encompass_Preferred
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA poly
Catalog No. 50-184-069 Supplier Biochain Institute Inc Supplier No. R123520650
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA poly