Learn More
Biochain Institute Inc Total RNA - Human Tumor Tissue: Adrenal, 10 ug/PK
SDP
Catalog No. 50184056
Encompass_Preferred
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA poly
Catalog No. 50-184-056 Supplier Biochain Institute Inc Supplier No. R123500410
BioChain's Total RNAs are isolated from a wide variety of documented human normal, diseased, and tumor tissues, mouse, rat, monkey, and plant tissues. Total RNA isolation is performed using proprietary techniques. Contamination by RNase, genomic DNA poly