Learn More
Biochain Institute Inc Total RNA - Human Normal Lymphocyte Cell Line, Ambient temperature shipping, 100 ug/pk
SDP
Catalog No. 501871510
Encompass_Preferred
Total RNA is isolated by modified guanidine thiocyanate techniques and stored in RNA storage buffer. The vials were then dried for room temperature shipping and storage. The total RNA need to be reconsitituted before use. Store the vials at -80 ℃ after reconstitution.
Catalog No. 50-187-1510 Supplier Biochain Institute Inc Supplier No. ATR12541481
Total RNA is isolated by modified guanidine thiocyanate techniques and stored in RNA storage buffer. The vials were then dried for room temperature shipping and storage. The total RNA need to be reconsitituted before use. Store the vials at -80 ℃ after reconstitution.