Learn More
Norgen Biotek Corp Total RNA Purification Plus Kit, 100 Prep
Supplier:  Norgen Biotek Corp 48400
Encompass_Preferred
This kit purifies total RNA including miRNA from biological samples and employs an extra column for the efficient removal of contaminating gDNA, thereby replacing the enzymantic DNase step. Efficiently extract total RNA from a range of samples including cells, bacteria, yeast, virus and bodily fluids including plasma/serum, blood, saliva, CSF and more. Extract high quality and purity RNA with excellent RIN values and A260/A280 suitable for downstream applications including qRT-PCR, RT-PCR, microarrays, NGS and more.The kit purifies all sizes of RNA from large mRNA, lncRNA down to microRNA (miRNA) in the same fraction without the requirement of phenol. Isolate all RNA sequences at an equal rate irrespective of size. Moreover, when the RNA sequences are small (e.g. miRNA), the column binds small RNAs regardless of their GC content.
Catalog No. NC1598593