Learn More
Norgen Biotek Corp Total RNA Purification Plus Micro Kit
Supplier:  Norgen Biotek Corp 48500
Encompass_Preferred
Norgen’s Total RNA Purification Plus Micro Kit provides a rapid and sensitive method for the isolation and purification of total RNA from small input amounts of cultured animal cells, tissue samples, and microdissected samples including laser-capture microdissection (LCM). The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA). Genomic DNA is removed from the sample using a Genomic DNA Removal column, and the RNA is preferentially purified from other cellular components such as proteins, without the use of phenol or chloroform. The purified RNA is of the highest integrity, and can be used in a number of downstream applications including real time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays.
Catalog No. NC1304915