Learn More
Norgen Biotek Corp Total RNA Purification 96-Well Kit
Catalog No. NC0770399
Encompass_Preferred
This 96-well kit provides a rapid method for the high-throughput isolation and purification of total RNA in 30 minutes using vacuum manifold, plate centrifuge or liquid handlers with vacuum capabilities. Total RNA can be isolated from a broad range of sample sources including cultured cells, tissues, blood, serum, plasma, bacteria, yeast, fungi, and viruses. The eluted RNA is consistent from well-to-well with high quality and purity, with excellent RIN values and A260/A280, and is suitable for downstream applications including qRT-PCR, RT-PCR, microarrays, NGS and more. The kit purifies all sizes of RNA from large mRNA, lncRNA down to microRNA (miRNA) in the same fraction without the requirement of phenol. Isolate all RNA sequences at an equal rate irrespective of size. Moreover, when the RNA sequences are small (e.g. miRNA), the column binds small RNAs regardless of their GC content.
Catalog No. NC0770399 Supplier Norgen Biotek Corp Supplier No. 24300
This 96-well kit provides a rapid method for the high-throughput isolation and purification of total RNA in 30 minutes using vacuum manifold, plate centrifuge or liquid handlers with vacuum capabilities. Total RNA can be isolated from a broad range of sample sources including cultured cells, tissues, blood, serum, plasma, bacteria, yeast, fungi, and viruses. The eluted RNA is consistent from well-to-well with high quality and purity, with excellent RIN values and A260/A280, and is suitable for downstream applications including qRT-PCR, RT-PCR, microarrays, NGS and more. The kit purifies all sizes of RNA from large mRNA, lncRNA down to microRNA (miRNA) in the same fraction without the requirement of phenol. Isolate all RNA sequences at an equal rate irrespective of size. Moreover, when the RNA sequences are small (e.g. miRNA), the column binds small RNAs regardless of their GC content.