Learn More
STEMCELL Technologies Total RNA Purification Kit, Size: 50 Preparations
SDP
Catalog No. 501976799
Encompass
The Total RNA Purification Kit uses a membrane-based system to quickly purify intact total RNA from 1 x 10^2 to 5 x 10^6 cells without an organic extraction step, eliminating the need for additional purification steps. The Total RNA Purification Kit includes an on-column DNase I treatment to significantly reduce genomic DNA contamination and ensure the purified RNA is suitable for sensitive downstream applications. Intact total RNA can be isolated in as little as 30 minutes. The Total RNA Purification Kit is not recommended for the purification of small RNAs (< 200 nt), such as miRNA.
Catalog No. 50-197-6799 Supplier STEMCELL Technologies Supplier No. 79040
The Total RNA Purification Kit uses a membrane-based system to quickly purify intact total RNA from 1 x 10^2 to 5 x 10^6 cells without an organic extraction step, eliminating the need for additional purification steps. The Total RNA Purification Kit includes an on-column DNase I treatment to significantly reduce genomic DNA contamination and ensure the purified RNA is suitable for sensitive downstream applications. Intact total RNA can be isolated in as little as 30 minutes. The Total RNA Purification Kit is not recommended for the purification of small RNAs (< 200 nt), such as miRNA.