Learn More
Meso Scale Discovery Total S6RP WCL20ate
SDP
Catalog No. 5000080528
Encompass
The Total S6RP Whole Cell Lysate Kit from Mesoscale Discovery is designed for the detection of total S6 ribosomal protein and phospho-S6 ribosomal protein in cell lysates This electrochemiluminescence immunoassay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG and cell lysis and wash buffers The kit can quantitatively measure total and phosphorylated S6RP in cell lysates with high sensitivity and without the need for radioactive reagents It is suitable for studying mTOR pathway biology and screening compounds that modulate S6RP phosphorylation
Catalog No. 50-000-80528 Supplier Meso Scale Discovery Supplier No. K150DHD3
Add to Cart
Add to Cart
Request a Quote
The Total S6RP Whole Cell Lysate Kit from Mesoscale Discovery is designed for the detection of total S6 ribosomal protein and phospho-S6 ribosomal protein in cell lysates This electrochemiluminescence immunoassay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG and cell lysis and wash buffers The kit can quantitatively measure total and phosphorylated S6RP in cell lysates with high sensitivity and without the need for radioactive reagents It is suitable for studying mTOR pathway biology and screening compounds that modulate S6RP phosphorylation