Learn More
Meso Scale Discovery Total S6RP Whole Cell Lysate
SDP
Catalog No. 5000078255
Encompass
The Meso Scale Discovery S6RP is a 96-well plate coated with streptavidin for use in multiplex immunoassays It has high binding capacity and low non-specific binding The plate is compatible with MSD s electrochemiluminescence detection technology for sensitive and rapid quantification of biomarkers With its high-sensitivity and broad dynamic range the S6RP plate enables simultaneous detection of up to 10 biomarkers per well from a variety of sample types including serum plasma and cell culture supernatants The plate is suitable for sandwich immunoassays using MSD SULFO-TAG labels
Catalog No. 50-000-78255 Supplier Meso Scale Discovery Supplier No. K150DHD1
The Meso Scale Discovery S6RP is a 96-well plate coated with streptavidin for use in multiplex immunoassays It has high binding capacity and low non-specific binding The plate is compatible with MSD s electrochemiluminescence detection technology for sensitive and rapid quantification of biomarkers With its high-sensitivity and broad dynamic range the S6RP plate enables simultaneous detection of up to 10 biomarkers per well from a variety of sample types including serum plasma and cell culture supernatants The plate is suitable for sandwich immunoassays using MSD SULFO-TAG labels