Learn More
Meso Scale Discovery Total S6RP Whole Cell Lysate
SDP
Catalog No. 5000081829
Encompass
The Meso Scale Discovery S6RP is a highly sensitive electrochemiluminescent immunoassay kit for the quantitative detection of soluble receptor activator of nuclear factor kappa-B ligand (sRANKL) in human serum plasma and cell culture supernatants The assay utilizes MSD s patented SULFO-TAG labels and provides superior sensitivity and a broad dynamic range The kit includes a 96-well plate coated with an anti-sRANKL capture antibody a SULFO-TAG labeled anti-sRANKL detection antibody assay and sample diluents calibrators and a 10X read buffer The assay can detect sRANKL in the range of 0 08 - 500 pg/mL and requires only 50 L of sample
Catalog No. 50-000-81829 Supplier Meso Scale Discovery Supplier No. K150DHD2
The Meso Scale Discovery S6RP is a highly sensitive electrochemiluminescent immunoassay kit for the quantitative detection of soluble receptor activator of nuclear factor kappa-B ligand (sRANKL) in human serum plasma and cell culture supernatants The assay utilizes MSD s patented SULFO-TAG labels and provides superior sensitivity and a broad dynamic range The kit includes a 96-well plate coated with an anti-sRANKL capture antibody a SULFO-TAG labeled anti-sRANKL detection antibody assay and sample diluents calibrators and a 10X read buffer The assay can detect sRANKL in the range of 0 08 - 500 pg/mL and requires only 50 L of sample