Learn More
Cell Signaling Technology Matched Antibody Pair, Total SMAD4, Unconjugated, 1 Kit
SDP
Catalog No. 503134378
Encompass
The Total SMAD4 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom labeling capabilities. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and are provided in a BSA and azide-free formulation, facilitating conjugation with labels such as fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Features capture and detection antibodies validated for reactivity in human, mouse, rat, and monkey models.
  • Supplied in 1X PBS (pH 7.8).
  • BSA and azide-free composition supports direct conjugation to specialized labels.
  • Optimal for sensitive detection in various ELISA-based applications.
Catalog No. 50-313-4378 Supplier Cell Signaling Technology Supplier No. 45319S
The Total SMAD4 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with custom labeling capabilities. This pair includes capture and detection antibodies that bind to non-overlapping epitopes and are provided in a BSA and azide-free formulation, facilitating conjugation with labels such as fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Features capture and detection antibodies validated for reactivity in human, mouse, rat, and monkey models.
  • Supplied in 1X PBS (pH 7.8).
  • BSA and azide-free composition supports direct conjugation to specialized labels.
  • Optimal for sensitive detection in various ELISA-based applications.