Learn More
Cell Signaling Technology Total Sox2 Matched Antibody Pair, Unconjugated, 1 Kit
SDP
Catalog No. 503133225
Encompass
The Total Sox2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms requiring pairs with custom antibody labeling, including fluorophores, lanthanides, biotin, and beads. The pair is compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Consists of capture and detection antibodies binding to non-overlapping epitopes
  • Validated for ELISA+ applications
  • Species reactivity: Human
Catalog No. 50-313-3225 Supplier Cell Signaling Technology Supplier No. 55220S
The Total Sox2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms requiring pairs with custom antibody labeling, including fluorophores, lanthanides, biotin, and beads. The pair is compatible with platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Consists of capture and detection antibodies binding to non-overlapping epitopes
  • Validated for ELISA+ applications
  • Species reactivity: Human