Learn More
Cell Signaling Technology Matched Antibody Pair, Total Src, Human and Mouse Reactivity, 1 Kit
SDP
Catalog No. 503133991
Encompass
The Total Src Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling, such as fluorophores, lanthanides, biotin, or beads. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes 100 μg each of rabbit and mouse monoclonal antibodies
  • Optimized for ELISA applications
  • Supplied in 1X PBS (pH 7.8), BSA and azide free
  • Compatible with Human and Mouse species
  • Requires storage at -20°C
Catalog No. 50-313-3991 Supplier Cell Signaling Technology Supplier No. 92214S
The Total Src Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling, such as fluorophores, lanthanides, biotin, or beads. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes 100 μg each of rabbit and mouse monoclonal antibodies
  • Optimized for ELISA applications
  • Supplied in 1X PBS (pH 7.8), BSA and azide free
  • Compatible with Human and Mouse species
  • Requires storage at -20°C