Learn More
Meso Scale Discovery Total STAT3 Kit, 25 plates, 96-well format
SDP
Catalog No. NC3254279
Encompass
The MESO SCALE DISCOVERY Total STAT3 Kit is designed for the quantitative detection of total Signal Transducer and Activator of Transcription 3 (STAT3) protein in cellular lysates. This kit is ideal for researchers studying intracellular signaling pathways and phosphoprotein analysis, validated across human, mouse, and rat species. With its 25-plate format, it supports high-throughput applications, ensuring reliable results while streamlining experimental setups with all necessary components included.
  • Quantitative detection of total STAT3 protein levels
  • Validated for human, mouse, and rat samples
  • 25-plate kit for larger studies and screening applications
  • All necessary components included for a complete assay
  • Compatible with multiple MSD instruments for high sensitivity
Catalog No. NC3254279 Supplier Meso Scale Discovery Supplier No. K150SND4
Add to Cart
Add to Cart
Request a Quote
The MESO SCALE DISCOVERY Total STAT3 Kit is designed for the quantitative detection of total Signal Transducer and Activator of Transcription 3 (STAT3) protein in cellular lysates. This kit is ideal for researchers studying intracellular signaling pathways and phosphoprotein analysis, validated across human, mouse, and rat species. With its 25-plate format, it supports high-throughput applications, ensuring reliable results while streamlining experimental setups with all necessary components included.
  • Quantitative detection of total STAT3 protein levels
  • Validated for human, mouse, and rat samples
  • 25-plate kit for larger studies and screening applications
  • All necessary components included for a complete assay
  • Compatible with multiple MSD instruments for high sensitivity