Learn More
Meso Scale Discovery Total STAT3 Kit
SDP
Catalog No. 5000078367
Encompass
The Total STAT3 Kit from Meso Scale Discovery is a 96-well plate assay for measuring total STAT3 protein in cell and tissue lysates It uses electrochemiluminescence technology for high sensitivity and a broad dynamic range The assay takes 3 5 hours with 2 5 hours of hands-on time The kit includes a STAT3 standard detection antibody Read Buffer and 96-well plates with capture antibody It performs up to 40 tests in duplicate with a detection range of 50-50 000 pg/mL and minimal cross-reactivity with other STAT proteins providing a fast sensitive and robust assay
Catalog No. 50-000-78367 Supplier Meso Scale Discovery Supplier No. K150SND2
The Total STAT3 Kit from Meso Scale Discovery is a 96-well plate assay for measuring total STAT3 protein in cell and tissue lysates It uses electrochemiluminescence technology for high sensitivity and a broad dynamic range The assay takes 3 5 hours with 2 5 hours of hands-on time The kit includes a STAT3 standard detection antibody Read Buffer and 96-well plates with capture antibody It performs up to 40 tests in duplicate with a detection range of 50-50 000 pg/mL and minimal cross-reactivity with other STAT proteins providing a fast sensitive and robust assay