Learn More
Cell Signaling Technology Total Stat5a Matched Antibody Pair, Unconjugated, Human, ELISA, 1 Kit
SDP
Catalog No. 503131766
Encompass
The Total Stat5a Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms, including MSD, Quanterix Simoa, Alpha Technology, and Luminex. This set includes capture and detection antibodies that bind to non-overlapping epitopes and is ideal for projects requiring specialized or custom labeling with fluorophores, lanthanides, biotin, or beads.
  • Recognizes Stat5a without cross-reacting with Stat5b
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • Free of BSA and azide
  • Validated for human species reactivity
  • Components consist of mouse monoclonal capture antibody and rabbit monoclonal detection antibody
Catalog No. 50-313-1766 Supplier Cell Signaling Technology Supplier No. 36289S
The Total Stat5a Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms, including MSD, Quanterix Simoa, Alpha Technology, and Luminex. This set includes capture and detection antibodies that bind to non-overlapping epitopes and is ideal for projects requiring specialized or custom labeling with fluorophores, lanthanides, biotin, or beads.
  • Recognizes Stat5a without cross-reacting with Stat5b
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • Free of BSA and azide
  • Validated for human species reactivity
  • Components consist of mouse monoclonal capture antibody and rabbit monoclonal detection antibody