Learn More
Meso Scale Discovery Total STAT5ab WCL
SDP
Catalog No. 5000081599
Encompass
The Total STAT5A/B Whole Cell Lysate Kit from Meso Scale Discovery is used to detect total STAT5A/B protein in cell lysates The kit contains a 96-well plate coated with capture antibody detection antibody read buffer sample diluent wash buffer and lyophilized STAT5A/B standard The electrochemiluminescent assay allows for the quantitative measurement of total STAT5A/B with high sensitivity and a broad dynamic range The kit can process up to 80 samples in duplicate and results are obtained in just 3 5 hours
Catalog No. 50-000-81599 Supplier Meso Scale Discovery Supplier No. K150IHD1
The Total STAT5A/B Whole Cell Lysate Kit from Meso Scale Discovery is used to detect total STAT5A/B protein in cell lysates The kit contains a 96-well plate coated with capture antibody detection antibody read buffer sample diluent wash buffer and lyophilized STAT5A/B standard The electrochemiluminescent assay allows for the quantitative measurement of total STAT5A/B with high sensitivity and a broad dynamic range The kit can process up to 80 samples in duplicate and results are obtained in just 3 5 hours