Learn More
Meso Scale Discovery Total STAT5ab WCL20ate
SDP
Catalog No. 5000078819
Encompass
The Total STAT5A/B Whole Cell Lysate Kit from Meso Scale Discovery is used to measure STAT5A/B protein levels in cell lysates The kit contains a 96-well plate coated with capture antibody a detection antibody and buffers and reagents needed to perform the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range The kit can be used to monitor STAT5A/B activation and expression levels in response to cytokines growth factors and drugs that modulate the JAK/STAT pathway With a sample incubation time of just 2 hours this kit enables rapid analysis of STAT5A/B levels across multiple samples
Catalog No. 50-000-78819 Supplier Meso Scale Discovery Supplier No. K150IHD3
Add to Cart
Add to Cart
Request a Quote
The Total STAT5A/B Whole Cell Lysate Kit from Meso Scale Discovery is used to measure STAT5A/B protein levels in cell lysates The kit contains a 96-well plate coated with capture antibody a detection antibody and buffers and reagents needed to perform the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range The kit can be used to monitor STAT5A/B activation and expression levels in response to cytokines growth factors and drugs that modulate the JAK/STAT pathway With a sample incubation time of just 2 hours this kit enables rapid analysis of STAT5A/B levels across multiple samples