Learn More
Cell Signaling Technology Total Survivin Matched Antibody Pair, Unconjugated, Human and Monkey Reactivity, 1 Kit
SDP
Catalog No. 503134021
Encompass
The Total Survivin Matched Antibody Pair is designed for use in immunoassay technologies, including high-throughput ELISA platforms, that require capture and detection antibodies binding to non-overlapping epitopes. This kit is suitable for applications requiring custom antibody labeling, such as with fluorophores, lanthanides, biotin, or beads, and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes 100 μg each of mouse monoclonal and rabbit monoclonal antibodies
  • Supplied in 1X PBS buffer at pH 7.8, BSA and azide free
  • Validated for reactivity in human and monkey samples
  • Optimized for ELISA and ELISA-like assays
  • Store at -20°C; aliquot into single-use vials to avoid repeated freeze/thaw cycles
Catalog No. 50-313-4021 Supplier Cell Signaling Technology Supplier No. 87112S
The Total Survivin Matched Antibody Pair is designed for use in immunoassay technologies, including high-throughput ELISA platforms, that require capture and detection antibodies binding to non-overlapping epitopes. This kit is suitable for applications requiring custom antibody labeling, such as with fluorophores, lanthanides, biotin, or beads, and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes 100 μg each of mouse monoclonal and rabbit monoclonal antibodies
  • Supplied in 1X PBS buffer at pH 7.8, BSA and azide free
  • Validated for reactivity in human and monkey samples
  • Optimized for ELISA and ELISA-like assays
  • Store at -20°C; aliquot into single-use vials to avoid repeated freeze/thaw cycles