Learn More
Alpco Ltd Total Thyroxine ELISA (Total T4 ELISA)
SDP
Supplier:  Alpco Ltd 25TT4HUE01
Encompass_Preferred
The Total Thyroxine ELISA (Total T4 ELISA) is for the quantitative determination of the Total Thyroxine (T4) concentration in human Serum. For Research Use Only. Not For Use In Diagnostic Procedures.
Catalog No. NC2284568