Learn More
Innovative Research Inc Mouse Total Urokinase (uPA) ELISA Kit
Supplier:  Innovative Research Inc IMSUPAKTT
Encompass_Preferred
Mouse uPA total antigen assay is intended for the quantitative determination of total plasminogen activator antigen in mouse plasma. For research use only. Strip well format. Reagents for up to 96 tests.
Catalog No. 50-203-6145