Learn More
Cell Signaling Technology Total Vimentin Matched Antibody Pair, Human and Monkey Reactivity, Unconjugated, 1 Kit
SDP
Catalog No. 503132291
Encompass
The Total Vimentin Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require capture and detection antibodies with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This pair consists of antibodies that bind to non-overlapping epitopes and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as 100 μg of each capture and detection antibody
  • BSA and azide free formulation
  • Validated for reactivity in human and monkey species
  • Ideal for development of customized conjugate assays
  • Compatible with diverse high-throughput immunoassay platforms
  • Provided in 1X PBS (pH 7.8)
Catalog No. 50-313-2291 Supplier Cell Signaling Technology Supplier No. 28367S
The Total Vimentin Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require capture and detection antibodies with specialized or custom labeling, such as fluorophores, lanthanides, biotin, or beads. This pair consists of antibodies that bind to non-overlapping epitopes and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as 100 μg of each capture and detection antibody
  • BSA and azide free formulation
  • Validated for reactivity in human and monkey species
  • Ideal for development of customized conjugate assays
  • Compatible with diverse high-throughput immunoassay platforms
  • Provided in 1X PBS (pH 7.8)