Learn More
Cell Signaling Technology Total YAP Matched Antibody Pair, Human, Mouse, Unconjugated, 1 Kit
SDP
Catalog No. 503134022
Encompass
The Total YAP Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with custom labeling capabilities. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Validated for ELISA+ applications
  • Reacts with human and mouse proteins
  • Storage at -20°C recommended; aliquot into single-use vials to avoid multiple freeze/thaw cycles
Catalog No. 50-313-4022 Supplier Cell Signaling Technology Supplier No. 87446S
The Total YAP Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with custom labeling capabilities. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8))
  • BSA and azide free formulation
  • Validated for ELISA+ applications
  • Reacts with human and mouse proteins
  • Storage at -20°C recommended; aliquot into single-use vials to avoid multiple freeze/thaw cycles