Learn More
Cell Signaling Technology Matched Antibody Pair, ZAP70, Unconjugated, Human, Mouse, 1 Kit
SDP
Catalog No. 503132572
Encompass
The Total Zap-70 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs for specialized or custom labeling with fluorophores, lanthanides, biotin, or beads. These unconjugated antibodies are suitable for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in 1X PBS (pH 7.8) that is BSA and azide free.
  • Validated for reactivity in human and mouse samples.
  • Compatible with diverse assay platforms including AlphaScreen, AlphaLISA, LANCE, HTRF, and Luminex.
  • Optimized for research use only.
Catalog No. 50-313-2572 Supplier Cell Signaling Technology Supplier No. 28260S
The Total Zap-70 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs for specialized or custom labeling with fluorophores, lanthanides, biotin, or beads. These unconjugated antibodies are suitable for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Includes capture and detection antibodies that bind to non-overlapping epitopes.
  • Supplied in 1X PBS (pH 7.8) that is BSA and azide free.
  • Validated for reactivity in human and mouse samples.
  • Compatible with diverse assay platforms including AlphaScreen, AlphaLISA, LANCE, HTRF, and Luminex.
  • Optimized for research use only.