Learn More
SW Sustainability Solutions TotalForm TF-14BK General Purpose Bio-based Nitrile Gloves
GREENER_CHOICESDP
Catalog No. 17355294 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-294 X-Large
17-355-290 Small
17-355-289 Medium
17-355-288 Large
17-355-291 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-355-294 Supplier SW Sustainability Solutions Supplier No. C141005
Add to Cart
Add to Cart
General purpose nitrile gloves with liner made of 55% bio-based materials and EcoTek™ sustainable nitrile coating, ideal for laboratory applications.
Bio-based refers to the monomer materials to produce the polymer originating from natural sources.
  • EcoTek™ Sustainable Technology nitrile coating
  • Natural fiber liner is made of 55% Bio-based materials, sourced from natural resources and not petroleum-based
  • Flexible
  • Durable
  • High fit feel comfort
  • Impressive fingertip dexterity
  • Ideal dry grip performance
  • Touch screen compatibility
  • For use in laboratory applications
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Chlorinated No
Construction Knit
Coating Material Nitrile
Color Black, Gray
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nylon, Spandex
Product Type Eco-friendly Work Glove
Size X-Large
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Thumb Style Straight
Show More Show Less
*EcoTek gloves tested to ASTM D5526-12 Standard Test Method for Determining Anaerobic Biodegradation of Plastic Materials Under Accelerated Landfill Conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated.