Learn More
Andwin Scientific TOTALLY TUBULAR RACK SYSTEM
SDP
Catalog No. 50819941
Encompass_Preferred
The Totally Tubular modular rack system can accommodate four different sizes of tubes. The rack is recommended for use of storing 15 and 50 ml conical tubes. It can also hold 1.5 and 2.0 ml microcentrifuge tubes; and 12 x 75, 13 x 100, 16 x 125, and 17 x 100 mm culture tubes. This product is autoclavable and freezable. It measures 9.5 L x 4 W x 3.75 H. It is sold in the mixed neon colors of orange, green, pink, and yellow.
Catalog No. 50-819-941 Supplier Andwin Scientific Supplier No. 23441040
The Totally Tubular modular rack system can accommodate four different sizes of tubes. The rack is recommended for use of storing 15 and 50 ml conical tubes. It can also hold 1.5 and 2.0 ml microcentrifuge tubes; and 12 x 75, 13 x 100, 16 x 125, and 17 x 100 mm culture tubes. This product is autoclavable and freezable. It measures 9.5 L x 4 W x 3.75 H. It is sold in the mixed neon colors of orange, green, pink, and yellow.