Learn More
Andwin Scientific TOTALLY TUBULAR RACK SYSTEM
SDP
Supplier:  Andwin Scientific 23441000
Encompass_Preferred
The Totally Tubular modular rack system can accommodate four different sizes of tubes. The rack is recommended for use of storing 15 and 50 ml conical tubes. It can also hold 1.5 and 2.0 ml microcentrifuge tubes; and 12 x 75, 13 x 100, 16 x 125, and 17 x 100 mm culture tubes. This product is autoclavable and freezable. It measures 9.5 L x 4 W x 3.75 H. It is sold in the mixed standard colors of blue, green, red, and yellow.
Catalog No. 50-819-940