Learn More
Biolegend TotalSeq™-A0024 anti-human CD112 (Nectin-2)
Supplier:  Biolegend 337417
Encompass_Preferred
TotalSeq™-A0024 anti-human CD112 (Nectin-2) [TX31]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-6281