Learn More
Biolegend TotalSeq™-A0028 anti-human CD30
Supplier:  Biolegend 333913
Encompass_Preferred
TotalSeq™-A0028 anti-human CD30 [BY88]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-6142