Learn More
Biolegend TotalSeq™-A0046 anti-human CD8
Supplier:  Biolegend 344751
Encompass_Preferred
TotalSeq™-A0046 anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: African Green, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: PG; Size: 10 ug
Catalog No. NC1668124