Learn More
Biolegend TotalSeq™-A0084 anti-human CD56 (NCAM) Recombinant
Supplier:  Biolegend 392421
Encompass_Preferred
TotalSeq™-A0084 anti-human CD56 (NCAM) Recombinant [QA17A16]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-8910