Learn More
Biolegend TotalSeq™-A0356 anti-human CD254 (TRANCE, RANKL)
Supplier:  Biolegend 347509
Encompass_Preferred
TotalSeq™-A0356 anti-human CD254 (TRANCE, RANKL) [MIH24]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-6835