Learn More
Biolegend TotalSeq™-A0370 anti-human CD303 (BDCA-2)
Supplier:  Biolegend 354239
Encompass_Preferred
TotalSeq™-A0370 anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-7344