Learn More
Biolegend TotalSeq™-A0446 anti-human CD93
Supplier:  Biolegend 336121
Encompass_Preferred
TotalSeq™-A0446 anti-human CD93 [VIMD2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-6207