Learn More
Biolegend TotalSeq™-A0819 anti-human CD126 (IL-6Rα)
Supplier:  Biolegend 352813
Encompass_Preferred
TotalSeq™-A0819 anti-human CD126 (IL-6Rα) [UV4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-7151