Learn More
Biolegend TotalSeq™-A0901 anti-human GARP (LRRC32)
Supplier:  Biolegend 352515
Encompass_Preferred
TotalSeq™-A0901 anti-human GARP (LRRC32) [7B11]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-7141