Learn More
Biolegend TotalSeq™-B0026 anti-human CD47
Supplier:  Biolegend 323135
Encompass_Preferred
TotalSeq™-B0026 anti-human CD47 [CC2C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-5591