Learn More
Biolegend TotalSeq™-B0143 anti-human CD196 (CCR6)
Supplier:  Biolegend 353445
Encompass_Preferred
TotalSeq™-B0143 anti-human CD196 (CCR6) [G034E3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: PG; Size: 10 ug
Catalog No. 50-207-7262