Learn More
Biolegend TotalSeq™-B0161 anti-human CD11b
Supplier:  Biolegend 301357
Encompass_Preferred
TotalSeq™-B0161 anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: PG; Size: 10 ug
Catalog No. NC1652783