Learn More
Biolegend TotalSeq™-B0252 anti-human Hashtag 2
Supplier:  Biolegend 394633
Encompass_Preferred
TotalSeq™-B0252 anti-human Hashtag 2 [LNH-94; 2M2]; Isotype: Mouse IgG1, κ (all clones); Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-9122