Learn More
Biolegend TotalSeq™-B0394 anti-human CD71
Supplier:  Biolegend 334127
Encompass_Preferred
TotalSeq™-B0394 anti-human CD71 [CY1G4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. 50-207-6159