Learn More
Biolegend TotalSeq™-B0817 anti-human CD109
Supplier:  Biolegend 323311
Encompass_Preferred
TotalSeq™-B0817 anti-human CD109 [W7C5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Cynomolgus; Apps: PG; Size: 10 ug
Catalog No. 50-207-5603