Learn More
Biolegend TotalSeq™-B0912 anti-human GPR56
Supplier:  Biolegend 358211
Encompass_Preferred
TotalSeq™-B0912 anti-human GPR56 [CG4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: PG; Size: 10 ug
Catalog No. 50-213-1955